missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%

CAS: 764-01-2 | C4H6O | 70.091 g/mol

Supplier:  Thermo Scientific Chemicals A1570914

 View more versions of this product

Catalog No. AAA1570914

Only null left
Explore available promotions

Description

Description

2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
Specifications

Specifications

2-Butyn-1-ol
-2°C
142°C to 143°C
Characteristic
1.453
25 g
1733676
Miscible with chloroform and methanol.
CC#CCO
70.091
70.09
764-01-2
0.938
51°C (123°F)
C4H6O
MFCD00002914
UN1987
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
NEEDEQSZOUAJMU-UHFFFAOYSA-N
but-2-yn-1-ol
12991
98%

Chemical Identifiers

764-01-2
70.091
NEEDEQSZOUAJMU-UHFFFAOYSA-N
12991
CC#CCO
C4H6O
MFCD00002914
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
but-2-yn-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 212-113-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only