missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%

Catalog No. p-80766
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223400050 5 mL Glass bottle
AC223400250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223400050 Supplier Thermo Scientific Chemicals Supplier No. 223400050
Only null left

CAS: 764-01-2 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 764-01-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00002914
InChI Key NEEDEQSZOUAJMU-UHFFFAOYSA-N
Synonym 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
PubChem CID 12991
IUPAC Name but-2-yn-1-ol
SMILES CC#CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.2°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 142.0°C to 143.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Linear Formula CH3C≡CCH2OH
Quantity 5 mL
Beilstein 01, II, 505
Specific Gravity 0.93
Formula Weight 70.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butyn-1-ol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: R
EINECSNumber 212-113-4
TSCA TSCA

RUO – Research Use Only