missing translation for 'onlineSavingsMsg'
Learn More

2-(Butylthio)ethanol 98.0+%, TCI America™

Catalog No. b09175ml
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
B09175ML 5 mL
1 options

Catalog No. B09175ML

Supplier: TCI America B09175ML

Only null left

Chemical Identifiers

CAS 5331-37-3
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.24
MDL Number MFCD00014042
InChI Key ARRJJHJQSNPJFV-UHFFFAOYSA-N
Synonym Butyl 2-Hydroxyethyl Sulfide
PubChem CID 79241
IUPAC Name 2-(butylsulfanyl)ethan-1-ol
SMILES CCCCSCCO

Specifications

Color Yellow
Boiling Point 217°C
Quantity 5 mL
Formula Weight 134.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Butylthio)ethanol