missing translation for 'onlineSavingsMsg'
Learn More

2-Butylimidazole 97.0+%, TCI America™

Catalog No. B31365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31365G 5 g
B313625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31365G Supplier TCI America Supplier No. B31365G

Chemical Identifiers

CAS 50790-93-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00792664
InChI Key SLLDUURXGMDOCY-UHFFFAOYSA-N
PubChem CID 11815351
IUPAC Name 2-butyl-1H-imidazole
SMILES CCCCC1=NC=CN1

Specifications

Melting Point 52°C
Boiling Point 294°C
Quantity 5 g
Formula Weight 124.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butylimidazole