missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-n-octan-1-amine 98.0+%, TCI America™

Catalog No. B55505ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55505ML 5 mL
B55501ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55505ML Supplier TCI America Supplier No. B55505ML
Only null left

Chemical Identifiers

CAS 217655-07-7
Molecular Formula C12H27N
Synonym 2-Butyl-1-n-octylamine, 1-Amino-2-butyl-n-octane, 5-(Aminomethyl)undecane

Specifications

Color Colorless
Quantity 5 mL
UN Number 2735
Formula Weight 185.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Butyl-n-octan-1-amine