missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butyl-1H-imidazole-5-carboxaldehyde 98.0+%, TCI America™
$356.03 - $1213.51
Chemical Identifiers
| CAS | 68282-49-5 |
|---|---|
| Molecular Formula | C8H12N2O |
| Molecular Weight (g/mol) | 152.197 |
| MDL Number | MFCD01075030 |
| InChI Key | PTHGVOCFAZSNNA-UHFFFAOYSA-N |
| Synonym | 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde |
| PubChem CID | 2735671 |
| IUPAC Name | 2-butyl-1H-imidazole-5-carbaldehyde |
| SMILES | CCCCC1=NC=C(N1)C=O |
Chemical Identifiers
| 68282-49-5 | |
| 152.197 | |
| PTHGVOCFAZSNNA-UHFFFAOYSA-N | |
| 2735671 | |
| CCCCC1=NC=C(N1)C=O |
| C8H12N2O | |
| MFCD01075030 | |
| 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde | |
| 2-butyl-1H-imidazole-5-carbaldehyde |
Specifications
| 68282-49-5 | |
| Yellow | |
| MFCD01075030 | |
| 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde | |
| CCCCC1=NC=C(N1)C=O | |
| 152.197 | |
| 152.20 | |
| Crystalline Powder |
| 118°C | |
| C8H12N2O | |
| 5 g | |
| PTHGVOCFAZSNNA-UHFFFAOYSA-N | |
| 2-butyl-1H-imidazole-5-carbaldehyde | |
| 2735671 | |
| ≥98.0% (GC) | |
| 2-Butyl-1H-imidazole-5-carboxaldehyde |
Safety and Handling
EINECSNumber : (5)-6676
TSCA : No