missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-1H-imidazole-5-carboxaldehyde 98.0+%, TCI America™

Catalog No. B40645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40645G 5 g
B406425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40645G Supplier TCI America Supplier No. B40645G
Only null left

Chemical Identifiers

CAS 68282-49-5
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD01075030
InChI Key PTHGVOCFAZSNNA-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
PubChem CID 2735671
IUPAC Name 2-butyl-1H-imidazole-5-carbaldehyde
SMILES CCCCC1=NC=C(N1)C=O

Specifications

Melting Point 118°C
Color Yellow
Quantity 5 g
Formula Weight 152.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-1H-imidazole-5-carboxaldehyde
EINECSNumber (5)-6676
TSCA No