missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 68282-49-5
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD01075030
InChI Key PTHGVOCFAZSNNA-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
PubChem CID 2735671
IUPAC Name 2-butyl-1H-imidazole-5-carbaldehyde
SMILES CCCCC1=NC=C(N1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SB02028DA
SDS View Documents
Thermo Scientific Chemicals
SB02028DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

68282-49-5
152.197
PTHGVOCFAZSNNA-UHFFFAOYSA-N
2735671
CCCCC1=NC=C(N1)C=O
C8H12N2O
MFCD01075030
2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
2-butyl-1H-imidazole-5-carbaldehyde

Specifications

68282-49-5
97%
C8H12N2O
1g
PTHGVOCFAZSNNA-UHFFFAOYSA-N
2-butyl-1H-imidazole-5-carbaldehyde
2735671
97%
97.0
Amber glass bottle
MFCD01075030
2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
CCCCC1=NC=C(N1)C=O
152.197
152.2
2-Butyl-1H-imidazole-4-carbaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'