missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butyl-1-n-octanol 98.0+%, TCI America™
Supplier: TCI America B3457100ML
Specifications
| 2-Butyl-1-n-octanol | |
| -80°C | |
| 104°C | |
| MFCD00053508 | |
| 3082 | |
| XMVBHZBLHNOQON-UHFFFAOYNA-N | |
| 2-butyloctan-1-ol | |
| 19800 | |
| 186.34 | |
| Liquid |
| 3913-02-8 | |
| Colorless | |
| C12H26O | |
| 100 mL | |
| 2-Butyl-n-octyl Alcohol | |
| CCCCCCC(CCCC)CO | |
| 186.34 | |
| CHEBI:84235 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3913-02-8 | |
| 186.34 | |
| XMVBHZBLHNOQON-UHFFFAOYNA-N | |
| 19800 | |
| 2-butyloctan-1-ol |
| C12H26O | |
| MFCD00053508 | |
| 2-Butyl-n-octyl Alcohol | |
| CHEBI:84235 | |
| CCCCCCC(CCCC)CO |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
RTECSNumber : RH0885000
TSCA : Yes