missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butyl-1-n-octanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -80°C |
| Color | Colorless |
| Boiling Point | 104°C |
| Quantity | 500 mL |
| UN Number | 3082 |
| Formula Weight | 186.34 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Butyl-1-n-octanol |
Safety and Handling
| EINECSNumber | (2)-0217&(2)-3704 |
| RTECSNumber | RH0885000 |
| TSCA | Yes |