missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-1-n-octanol 98.0+%, TCI America™

Catalog No. B345725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B345725ML 25 mL
B3457100ML 100 mL
B3457500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B345725ML Supplier TCI America Supplier No. B345725ML
Only null left

Chemical Identifiers

CAS 3913-02-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00053508
InChI Key XMVBHZBLHNOQON-UHFFFAOYNA-N
Synonym 2-Butyl-n-octyl Alcohol
PubChem CID 19800
ChEBI CHEBI:84235
IUPAC Name 2-butyloctan-1-ol
SMILES CCCCCCC(CCCC)CO

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 104°C
Quantity 25 mL
UN Number 3082
Formula Weight 186.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butyl-1-n-octanol
EINECSNumber (2)-0217&(2)-3704
RTECSNumber RH0885000
TSCA Yes