missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 194.04 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Butoxyphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |