missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether), Reagents

Catalog No. p-200020876
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337384 1 L
01337383 20 L
01337382 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337384 Supplier Labchem Inc Supplier No. C21562501C
Only null left

2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether), NIST Traceable, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym Ethylene Glycol Monobutyl Ether, Butyl Cellosolve
IUPAC Name 2-butoxyethan-1-ol
SMILES CCCCOCCO

Specifications

Boiling Range 171°C
Melting Point -70°C
Color Colorless
Boiling Point 171°C
Melting Range -70°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.9019
Physical Form Liquid
Chemical Name or Material 2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether)