missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethanol, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B117720LTMT

 View more versions of this product

Catalog No. 18608201


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

111-76-2
Steel Can
0.97
1
20 L
Reagent

Chemical Identifiers

111-76-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.