missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyacetic acid, 98%

Catalog No. AC257760010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257760010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257760010 Supplier Thermo Scientific Chemicals Supplier No. 257760010
Only null left

CAS: 2516-93-0 | C6H12O3 | 132.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2516-93-0
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.15
MDL Number MFCD00067044
InChI Key AJQOASGWDCBKCJ-UHFFFAOYSA-N
Synonym butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa
PubChem CID 41958
IUPAC Name 2-butoxyacetic acid
SMILES CCCCOCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270
Quantity 1 g
Specific Gravity 1.01
Formula Weight 132.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butoxyacetic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only