missing translation for 'onlineSavingsMsg'
Learn More

2-Buten-1-ylsuccinic Anhydride 98.0+%, TCI America™

Catalog No. p-7119158
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16135G 5 g
B161325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16135G Supplier TCI America Supplier No. B16135G
Only null left

Chemical Identifiers

CAS 7538-42-3
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD03423563
InChI Key YCYZOWYCOUNUTM-UHFFFAOYSA-N
Synonym 4-Hexene-1,2-dicarboxylic Anhydride
PubChem CID 548964
IUPAC Name 3-but-2-enyloxolane-2,5-dione
SMILES CC=CCC1CC(=O)OC1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Buten-1-ylsuccinic Anhydride