missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7157737
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600098 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18600098 Supplier Spectrum Chemical Mfg Cor Supplier No. M1260100ML
Only null left

M1260, 78-93-3, C4H8O

Spectrum™ Chemical 2-Butanone, Reagent, ACS

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

CAS Max % 100%
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.0025%
Titratable Acid 0.0005 meq/g
UN Number UN1193
Water 0.2%
Boiling Point 80 °C
Percent Purity 99%
Grade Reagent
Chemical Name or Material 2-Butanone
Show More Show Less