missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone Oxime 99.0+%, TCI America™

Catalog No. B068825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068825ML 25 mL
B0688500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B068825ML Supplier TCI America Supplier No. B068825ML

Chemical Identifiers

CAS 96-29-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00013935
InChI Key WHIVNJATOVLWBW-SNAWJCMRSA-N
Synonym mek-oxime, butanone oxime, methyl ethyl ketone oxime, 2-butanone oxime, ethyl methyl ketoxime, usaf ek-906, ethyl-methylketonoxim, unii-51yge935u9, 2-butanone, oxime, methyl ethyl ketoxime
PubChem CID 5324276
IUPAC Name (E)-N-(butan-2-ylidene)hydroxylamine
SMILES CC\C(C)=N\O

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 153°C
Quantity 25 mL
UN Number 2810
Formula Weight 87.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butanone Oxime
EINECSNumber (2)-0546
RTECSNumber EL9275000
TSCA Yes