missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone oxime, 99%

Catalog No. p-7044779
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08196AE 100 mL
AAL08196AP 500 mL
AAL08196AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL08196AE Supplier Thermo Scientific Chemicals Supplier No. L08196AE
Only null left

CAS: 96-29-7 | C4H9NO | 87.12 g/mol

2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

Solubility
Soluble in water at 20°C 114 g/L. Soluble in most organic solvents .

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.

Chemical Identifiers

CAS 96-29-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00013935
InChI Key WHIVNJATOVLWBW-SNAWJCMRSA-N
Synonym mek-oxime, butanone oxime, methyl ethyl ketone oxime, 2-butanone oxime, ethyl methyl ketoxime, usaf ek-906, ethyl-methylketonoxim, unii-51yge935u9, 2-butanone, oxime, methyl ethyl ketoxime
PubChem CID 5324276
IUPAC Name (NZ)-N-butan-2-ylidenehydroxylamine
SMILES CC\C(C)=N\O

Specifications

Melting Point -30°C
Density 0.921
Boiling Point 152°C to 153°C
Flash Point 62°C (144°F)
Refractive Index 1.443
Quantity 100 mL
Beilstein 1698241
Solubility Information Soluble in water at 20°C 114g/L. Soluble in most organic solvents .
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material 2-Butanone oxime
Show More Show Less
Hazard Category H312-H317-H318-H351
Hazard Statement GHS H Statement
H318-H351-H312-H317-H227
Causes serious eye damage.
Suspected of causing cancer.
Harmful in contact with skin.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P312-P333+P313-P363-P501c
EINECSNumber 202-496-6
RTECSNumber EL9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only