missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone [for Spectrophotometry], TCI America™

Catalog No. p-7128858
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E014025ML 25 mL
E0140500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E014025ML Supplier TCI America Supplier No. E014025ML
Only null left

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1193
Boiling Point 79°C
Formula Weight 72.11
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2-Butanone [for Spectrophotometry]
EINECSNumber (2)-0542
RTECSNumber EL6475000
TSCA Yes