missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen

Catalog No. p-7111863
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 4 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000112 100 mL Glass Bottle
6000113 1 L Glass Bottle
6000114 2.5 L Glass Bottle
60048235 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000112 Supplier Honeywell Inc Supplier No. 34861100ML
Only null left

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C (literature)
CAS Max % 100.0000%
Density 0.805 g/mL at 25 °C(lit.)
Quantity 100 mL
Flash Point −4°C
Packaging Glass Bottle
Linear Formula C2H5COCH3
UN Number UN1193
Beilstein 741880
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
Boiling Point 79°C to 81°C
Formula Weight 72.11
Percent Purity >99.7%
Grade HPLC
Chemical Name or Material 2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone
EINECSNumber 201-159-0
Recommended Storage Room Temp