missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS reagent, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 389430025

Catalog No. AC389430025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Butanone
0.8060g/mL
2.5L
Glass Bottle
0.0025 % max.
0.0005 meq/g max.
C2H5COCH3
0.42 mPa.s (15°C)
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11
ACS Reagent
78-93-3
80.0°C
−7°C
1.3780 to 1.3800
0.806
C4H8O
MFCD00011648
0.20% Max.
Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
CCC(C)=O
72.11
CHEBI:28398
≥99.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only