missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™

Catalog No. p-7120057
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046601 1 L Glass Bottle
60046606 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046601 Supplier Honeywell Inc Supplier No. 3604731L
Only null left

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Specifications

Melting Point -87°C
Density 0.805g/mL at 25°C
Quantity 1 L
Flash Point −3°C (26.6°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index n20/D 1.379
Boiling Point 80°C