missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS, 99+%

Maximum level of impurities: Color (APHA) 15, Residue after evaporation 0.0025%, Titratable acid 0.0005meq/g, H2O 0.20% | CAS: 78-93-3 | C4H8O | 72.11 g/mol

Supplier:  Thermo Scientific Chemicals 039119K2

 View more versions of this product

Catalog No. AA39119K2

Only null left
Explore available promotions

Description

Description

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Butanone
78-93-3
0.805
1 L
Liquid
99+%
MFCD00011648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11
°C to 87°C
Colorless
80°C
-3°C (26°F)
1.379
C4H8O
741880
Miscible in cold water,diethyl ether and acetone.
CCC(C)=O
72.11
CHEBI:28398
ACS

Chemical Identifiers

78-93-3
72.11
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
butan-2-one
C4H8O
MFCD00011648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CHEBI:28398
CCC(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336-H373

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 201-159-0

RTECSNumber : EL6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only