missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS, 99+%

Catalog No. AA39119M1
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39119M1 4 x 1 L
AA39119AP 500 mL
AA39119K2 1 L
AA39119K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA39119M1 Supplier Thermo Scientific Chemicals Supplier No. 039119M1

Maximum level of impurities: Color (APHA) 15, Residue after evaporation 0.0025%, Titratable acid 0.0005meq/g, H2O 0.20% | CAS: 78-93-3 | C4H8O | 72.11 g/mol

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point °C to 87°C
Color Colorless
Appearance Colorless liquid
Density 0.805
Quantity 4 x 1 L
Flash Point -3°C (26°F)
Ignition Point 514°C (957°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.379
Assay Percent Range >99%
Vapor Pressure 101 hPa (76mm Hg) at 20°C
Beilstein 741880
Boiling Point 80°C
Solubility Information Miscible in cold water,diethyl ether and acetone.
Formula Weight 72.11
Grade ACS
Chemical Name or Material 2-Butanone
Show More Show Less
Hazard Category H225-H319-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 201-159-0
RTECSNumber EL6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only