missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol 99.0+%, TCI America™

Supplier:  TCI America B0705500ML

 View more versions of this product

Catalog No. B0705500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Butanol
-115°C
100°C
MFCD00004569
1120
BTANRVKWQNVYAZ-UHFFFAOYNA-N
butan-2-ol
6568
74.12
Liquid
78-92-2
Colorless
C4H10O
500 mL
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
CCC(C)O
74.12
CHEBI:35687
≥99.0% (GC)

Chemical Identifiers

78-92-2
74.12
BTANRVKWQNVYAZ-UHFFFAOYNA-N
6568
butan-2-ol
C4H10O
MFCD00004569
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
CHEBI:35687
CCC(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3049

RTECSNumber : EO1750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions