missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -115°C |
| Color | Colorless |
| Density | 0.808g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.098 |
| Flash Point | 24°C (80.6°F) |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | n20/D 1.397 |
| Vapor Pressure | 12.5mmHg (20°C) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P305 + P351 + P338-P370 + P378 |
| Precautionary Statement | H226-H319-H335-H336 |
| DOTInformation | Class 3 - PG 3 - Butanols |
| EINECSNumber | 201-158-5 |
| RTECSNumber | EO1750000 |