missing translation for 'onlineSavingsMsg'
Learn More

2-Bromovaleric Acid 98.0+%, TCI America™

Catalog No. B066725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B066725G 25 g
B0667500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B066725G Supplier TCI America Supplier No. B066725G
Only null left

Chemical Identifiers

CAS 584-93-0
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00004217
InChI Key WMFATTFQNRPXBQ-UHFFFAOYNA-N
Synonym 2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
PubChem CID 11446
IUPAC Name 2-bromopentanoic acid
SMILES CCCC(Br)C(O)=O

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 g
UN Number 3265
Formula Weight 181.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromovaleric Acid