missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene 98.0+%, TCI America™

Catalog No. B094125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B094125G 25 g
B0941250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B094125G Supplier TCI America Supplier No. B094125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1003-09-4
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005417
InChI Key TUCRZHGAIRVWTI-UHFFFAOYSA-N
Synonym 2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
PubChem CID 13851
IUPAC Name 2-bromothiophene
SMILES BrC1=CC=CS1

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 g
UN Number 2929
Formula Weight 163.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromothiophene
EINECSNumber (9)-2166
TSCA Yes