missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole 98.0+%, TCI America™

Catalog No. b1280250g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
B1280250G 250 g
B128025G 25 g
2 options

Catalog No. B1280250G

Supplier: TCI America B1280250G

Only null left

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br

Specifications

Color Yellow
Boiling Point 171°C
Quantity 250 g
Formula Weight 164.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromothiazole
TSCA Yes
Recommended Storage Freezer