Learn More
2-Bromothiazole, 99%
CAS: 3034-53-5 | C3H2BrNS | 164.02 g/mol
Supplier: Thermo Scientific Chemicals A1483814
Description
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
Specifications
| 2-Bromothiazole | |
| 1.836 | |
| 63°C (145°F) | |
| 1.593 | |
| 25 g | |
| 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol | |
| RXNZFHIEDZEUQM-UHFFFAOYSA-N | |
| 2-bromo-1,3-thiazole | |
| 76430 | |
| 99% |
| 3034-53-5 | |
| 171°C | |
| C3H2BrNS | |
| MFCD00005316 | |
| 105724 | |
| It is insoluble in water. | |
| C1=CSC(=N1)Br | |
| 164.02 | |
| 164.02 |
Chemical Identifiers
| 3034-53-5 | |
| 164.02 | |
| RXNZFHIEDZEUQM-UHFFFAOYSA-N | |
| 76430 | |
| C1=CSC(=N1)Br |
| C3H2BrNS | |
| MFCD00005316 | |
| 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol | |
| 2-bromo-1,3-thiazole |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 221-229-4
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only