missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 99%

Catalog No. AAA1483809
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1483809 10 g
AAA1483814 25 g
AAA1483818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1483809 Supplier Thermo Scientific Chemicals Supplier No. A1483809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3034-53-5 | C3H2BrNS | 164.02 g/mol

2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br

Specifications

Density 1.836
Boiling Point 171°C
Flash Point 63°C (145°F)
Refractive Index 1.593
Quantity 10 g
Beilstein 105724
Solubility Information It is insoluble in water.
Formula Weight 164.02
Percent Purity 99%
Chemical Name or Material 2-Bromothiazole
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 221-229-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only