missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 98+%

CAS: 3034-53-5 | C3H2BrNS | 164.03 g/mol

$92.76 - $92.76

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.03
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC170910050
SDS View Documents
Thermo Scientific Chemicals
170910050
5 mL Glass bottle
Each for $92.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3034-53-5
164.03
RXNZFHIEDZEUQM-UHFFFAOYSA-N
76430
C1=CSC(=N1)Br
C3H2BrNS
MFCD00005316
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
2-bromo-1,3-thiazole

Specifications

3034-53-5
100.0
1.8200g/mL
63°C
98% min. (GC)
C3H2BrNS
MFCD00005316
27, 15
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
RXNZFHIEDZEUQM-UHFFFAOYSA-N
2-bromo-1,3-thiazole
76430
98+%
2-Bromothiazole, 98+%
98.0
Brown-Orange to Colorless
70°C to 71°C (20.0 mmHg)
Authentic
Glass bottle
1.5910 to 1.593
5 mL
1.82
Solubility in water: insoluble
C1=CSC(=N1)Br
164.03
164.03
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 221-229-4

TSCA : TSCA

Recommended Storage : Darkens on ageing

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only