missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 98+%

Catalog No. AC170910050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170910050 5 mL Glass bottle
AC170910250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170910050 Supplier Thermo Scientific Chemicals Supplier No. 170910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3034-53-5 | C3H2BrNS | 164.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.03
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Density 1.8200g/mL
Boiling Point 70°C to 71°C (20.0 mmHg)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5910 to 1.593
Quantity 5 mL
Beilstein 27, 15
Specific Gravity 1.82
Solubility Information Solubility in water: insoluble
Formula Weight 164.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Bromothiazole, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 221-229-4
TSCA TSCA
Recommended Storage Darkens on ageing

RUO – Research Use Only