missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromothiazole-5-carboxylic Acid 97.0+%, TCI America™
Supplier: TCI America B43295G
Specifications
| 2-Bromothiazole-5-carboxylic Acid | |
| 175°C | |
| C4H2BrNO2S | |
| 5 g | |
| BESGTWHUMYHYEQ-UHFFFAOYSA-N | |
| 2-bromo-1,3-thiazole-5-carboxylic acid | |
| 2763210 | |
| ≥97.0% (GC,T) |
| 54045-76-0 | |
| White-Yellow | |
| MFCD04115730 | |
| 2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic | |
| C1=C(SC(=N1)Br)C(=O)O | |
| 208.029 | |
| 208.03 | |
| Crystalline Powder |
Chemical Identifiers
| 54045-76-0 | |
| 208.029 | |
| BESGTWHUMYHYEQ-UHFFFAOYSA-N | |
| 2763210 | |
| C1=C(SC(=N1)Br)C(=O)O |
| C4H2BrNO2S | |
| MFCD04115730 | |
| 2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic | |
| 2-bromo-1,3-thiazole-5-carboxylic acid |
Safety and Handling
TSCA : No