missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole-5-carboxylic Acid 97.0+%, TCI America™

Catalog No. B43291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43291G 1 g
B43295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43291G Supplier TCI America Supplier No. B43291G

Chemical Identifiers

CAS 54045-76-0
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.029
MDL Number MFCD04115730
InChI Key BESGTWHUMYHYEQ-UHFFFAOYSA-N
Synonym 2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic
PubChem CID 2763210
IUPAC Name 2-bromo-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)Br)C(=O)O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.03
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromothiazole-5-carboxylic Acid