missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 54045-76-0
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.03
MDL Number MFCD04115730
InChI Key BESGTWHUMYHYEQ-UHFFFAOYSA-N
Synonym 2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic
PubChem CID 2763210
IUPAC Name 2-bromo-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)Br)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC429760010
SDS View Documents
Thermo Scientific Chemicals
429760010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429760050
SDS View Documents
Thermo Scientific Chemicals
429760050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

54045-76-0
208.03
BESGTWHUMYHYEQ-UHFFFAOYSA-N
2763210
C1=C(SC(=N1)Br)C(=O)O
C4H2BrNO2S
MFCD04115730
2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic
2-bromo-1,3-thiazole-5-carboxylic acid

Specifications

54045-76-0
100.0
96% min. (HPLC)
C4H2BrNO2S
1g
BESGTWHUMYHYEQ-UHFFFAOYSA-N
2-bromo-1,3-thiazole-5-carboxylic acid
2763210
97%
96.0
Authentic
Glass bottle
MFCD04115730
2-bromothiazole-5-carboxylic acid, 2-bromo-5-thiazolecarboxylic acid, 2-bromo-5-thiazolecarboxylicacid, 2-bromo-thiazole-5-carboxylic acid, 2-bromo-5-carboxythiazole, 5-thiazolecarboxylic acid, 2-bromo, 2-bromthiazol-5-carboxylic acid, pubchem15963, acmc-1ap3n, 2-bromothiazole-5-carboxylic
C1=C(SC(=N1)Br)C(=O)O
208.03
208.03
2-Bromothiazole-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only