missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoquinoline 98.0+%, TCI America™

Catalog No. B35241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35241G 1 g
B35245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35241G Supplier TCI America Supplier No. B35241G
Only null left

Chemical Identifiers

CAS 2005-43-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00234480
InChI Key QKJAZPHKNWSXDF-UHFFFAOYSA-N
Synonym quinoline, 2-bromo, bromoquinoline, pubchem21827, acmc-1cfcf, 2-bromoquinoline, ksc201o8j, 2-bromoquinoline 1g, abbypharma ap-12-10890
PubChem CID 2762756
IUPAC Name 2-bromoquinoline
SMILES BrC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 115°C
Quantity 1 g
Formula Weight 208.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoquinoline