missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine, 99%

Catalog No. AAA1324136
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324136 500 g
AAA1324114 25 g
AAA1324122 100 g
AAA1324130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1324136 Supplier Thermo Scientific Chemicals Supplier No. A1324136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-04-6 | C5H4BrN | 157.998 g/mol

2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

Solubility
Slightly miscible with water.

Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.

Chemical Identifiers

CAS 109-04-6
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 157.998
MDL Number MFCD00006219
InChI Key IMRWILPUOVGIMU-UHFFFAOYSA-N
Synonym o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
PubChem CID 7973
ChEBI CHEBI:51574
IUPAC Name 2-bromopyridine
SMILES C1=CC=NC(=C1)Br

Specifications

Density 1.625
Boiling Point 192°C to 194°C
Flash Point 97°C (207°F)
Odor Characteristic
Refractive Index 1.572
Quantity 500 g
UN Number UN2810
Beilstein 105789
Solubility Information Slightly miscible with water.
Formula Weight 158
Percent Purity 99%
Chemical Name or Material 2-Bromopyridine
Show More Show Less
Hazard Category H226-H301+H331-H310-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-641-6
RTECSNumber US3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only