missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-5-carboxaldehyde, 95%

CAS: 149806-06-4 | C6H4BrNO | 186.01 g/mol

Supplier:  Thermo Scientific Chemicals H6386706

 View more versions of this product

Catalog No. AAH6386706


Only null left
Explore available promotions

Description

Description

A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
Specifications

Specifications

2-Bromopyridine-5-carboxaldehyde
104°C to 110°C (decomposition)
C6H4BrNO
5 g
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
6-bromopyridine-3-carbaldehyde
11769234
95%
149806-06-4
Yellow
MFCD04115419
Air Sensitive
Soluble in dichloromethane,ether,ethyl acetate and methanol.
BrC1=CC=C(C=O)C=N1
186.01
186.01
Solid

Chemical Identifiers

149806-06-4
186.01
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
11769234
BrC1=CC=C(C=O)C=N1
C6H4BrNO
MFCD04115419
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
6-bromopyridine-3-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only