missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-5-carboxaldehyde, 95%

Catalog No. AAH6386703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6386703 1 g
AAH6386706 5 g
AAH6386714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6386703 Supplier Thermo Scientific Chemicals Supplier No. H6386703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 149806-06-4 | C6H4BrNO | 186.01 g/mol

A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.

Chemical Identifiers

CAS 149806-06-4
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.01
MDL Number MFCD04115419
InChI Key PVUKGNBRJFTFNJ-UHFFFAOYSA-N
Synonym 6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
PubChem CID 11769234
IUPAC Name 6-bromopyridine-3-carbaldehyde
SMILES BrC1=CC=C(C=O)C=N1

Specifications

Melting Point 104°C to 110°C (decomposition)
Color Yellow
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol.
Formula Weight 186.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromopyridine-5-carboxaldehyde
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only