missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromopyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$115.57 - $115.57
Specifications
| CAS | 452972-08-6 |
|---|---|
| Melting Point | 132°C |
| Color | White-Yellow |
| Molecular Formula | C5H5BBrNO2 |
| MDL Number | MFCD04113805 |
Specifications
| 452972-08-6 | |
| White-Yellow | |
| MFCD04113805 | |
| B(C1=C(N=CC=C1)Br)(O)O | |
| 201.814 | |
| 201.81 |
| 132°C | |
| C5H5BBrNO2 | |
| ODWRFKJNUJLAHM-UHFFFAOYSA-N | |
| (2-bromopyridin-3-yl)boronic acid | |
| 12067058 | |
| Crystalline Powder |