missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrazine 98.0+%, TCI America™

Catalog No. p-7123486
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43521G 1 g
B43525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43521G Supplier TCI America Supplier No. B43521G
Only null left

Chemical Identifiers

CAS 56423-63-3
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.986
MDL Number MFCD08275680
InChI Key WGFCNCNTGOFBBF-UHFFFAOYSA-N
Synonym pyrazine, bromo, bromopyrazine, pyrazine, 2-bromo, 5-bromopyrazine, brom-pyrazin, 2-bromanylpyrazine, pubchem18036, acmc-209lsg, 2-bromopyrazine, pyrazine, bromo-9ci
PubChem CID 642800
IUPAC Name 2-bromopyrazine
SMILES C1=CN=C(C=N1)Br

Specifications

Boiling Point 61°C
Quantity 1 g
Formula Weight 158.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromopyrazine
TSCA No
Recommended Storage Refrigerator