missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrazine, 95%

Catalog No. AC458160010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458160010 1 g
AC458160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458160010 Supplier Thermo Scientific Chemicals Supplier No. 458160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56423-63-3 | C4H3BrN2 | 159.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56423-63-3
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 159.99
InChI Key WGFCNCNTGOFBBF-UHFFFAOYSA-N
Synonym pyrazine, bromo, bromopyrazine, pyrazine, 2-bromo, 5-bromopyrazine, brom-pyrazin, 2-bromanylpyrazine, pubchem18036, acmc-209lsg, 2-bromopyrazine, pyrazine, bromo-9ci
PubChem CID 642800
IUPAC Name 2-bromopyrazine
SMILES C1=CN=C(C=N1)Br

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 159.99
Percent Purity 95%
Chemical Name or Material 2-Bromopyrazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only