missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropiophenone 96.0+%, TCI America™

Catalog No. B293325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B293325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B293325G Supplier TCI America Supplier No. B293325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2114-00-3
Boiling Point 137°C
Molecular Formula C9H9BrO
Quantity 25 g
MDL Number MFCD00000145
UN Number 2810
Synonym 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
InChI Key WPDWOCRJBPXJFM-UHFFFAOYSA-N
SMILES CC(C(=O)C1=CC=CC=C1)Br
IUPAC Name 2-bromo-1-phenylpropan-1-one
Molecular Weight (g/mol) 213.074
PubChem CID 16452
Formula Weight 213.07
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromopropiophenone
Show More Show Less
RTECSNumber UG8400000
TSCA Yes
Recommended Storage Refrigerator