Learn More
2-Bromopropiophenone, 96%
CAS: 2114-00-3 | C9H9BrO | 213.074 g/mol
Supplier: Thermo Scientific Chemicals A1066122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Bromopropiophenone | |
| 1.417 | |
| >110°C (230°F) | |
| C9H9BrO | |
| MFCD00000145 | |
| UN2927 | |
| 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone | |
| CC(C(=O)C1=CC=CC=C1)Br | |
| 213.074 | |
| 213.08 |
| 2114-00-3 | |
| 245°C to 250°C | |
| Pungent | |
| 1.572 | |
| 100 g | |
| 508550 | |
| WPDWOCRJBPXJFM-UHFFFAOYSA-N | |
| 2-bromo-1-phenylpropan-1-one | |
| 16452 | |
| 96% |
Chemical Identifiers
| 2114-00-3 | |
| 213.074 | |
| WPDWOCRJBPXJFM-UHFFFAOYSA-N | |
| 16452 | |
| CC(C(=O)C1=CC=CC=C1)Br |
| C9H9BrO | |
| MFCD00000145 | |
| 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone | |
| 2-bromo-1-phenylpropan-1-one |
Safety and Handling
GHS H Statement
H331-H314-H318
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 218-307-5
RTECSNumber : UG8400000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only