missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropiophenone, 96%

Catalog No. AAA1066114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1066114 25 g
AAA1066122 100 g
AAA1066136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1066114 Supplier Thermo Scientific Chemicals Supplier No. A1066114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2114-00-3 | C9H9BrO | 213.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2114-00-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000145
InChI Key WPDWOCRJBPXJFM-UHFFFAOYSA-N
Synonym 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
PubChem CID 16452
IUPAC Name 2-bromo-1-phenylpropan-1-one
SMILES CC(C(=O)C1=CC=CC=C1)Br

Specifications

Density 1.417
Boiling Point 245°C to 250°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.572
Quantity 25 g
UN Number UN2927
Beilstein 508550
Formula Weight 213.08
Percent Purity 96%
Chemical Name or Material 2-Bromopropiophenone
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-307-5
RTECSNumber UG8400000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only