missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropionyl Chloride 97.0+%, TCI America™

Catalog No. p-7123354
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B410725G 25 g
B4107100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B410725G Supplier TCI America Supplier No. B410725G
Only null left

Chemical Identifiers

CAS 7148-74-5
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000711
InChI Key OZGMODDEIHYPRY-UHFFFAOYNA-N
Synonym 2-bromopropionyl chloride, propanoyl chloride, 2-bromo, 2-bromopropionylchloride, 2-bromo-propionyl chloride, 2-brominepropionylchloride, alpha-bromopropionyl chloride, 2-bromopropionic acid chloride, sgqdjlpqbsmuh@, 2-bromopropanoylchloride, acmc-1cjhb
PubChem CID 97980
IUPAC Name 2-bromopropanoyl chloride
SMILES CC(Br)C(Cl)=O

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 g
UN Number 2920
Formula Weight 171.42
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Bromopropionyl Chloride
EINECSNumber (9)-1148&(9)-1184&(9)-1234
TSCA No