missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromophenylethynyl)trimethylsilane, 98%

Catalog No. p-7035679
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5340203 1 g
AAH5340206 5 g
AAH5340214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5340203 Supplier Thermo Scientific Chemicals Supplier No. H5340203

CAS: 38274-16-7 | C11H13BrSi | 253.21 g/mol

(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 38274-16-7
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01321391
InChI Key FABNGXSCLHXUOH-UHFFFAOYSA-N
Synonym 2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
PubChem CID 4547398
SMILES C[Si](C)(C)C#CC1=CC=CC=C1Br

Specifications

Boiling Point 60°C to 80°C (2mmHg)
Quantity 1 g
Flash Point >113°C (235°F)
Solubility Information Slightly soluble in water.
IUPAC Name [2-(2-bromophenyl)ethynyl]trimethylsilane
Formula Weight 253.21
Percent Purity 98%
Refractive Index 1.554
Density 1.19
Chemical Name or Material (2-Bromophenylethynyl)trimethylsilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only