missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenethylamine, 99%

Catalog No. p-3777676
Change view
Click to view available options
Quantity:
5 g
20 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350570050 5 g Glass Bottle
AC350570200 20 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC350570050 Supplier Thermo Scientific Chemicals Supplier No. 350570050

CAS: 65185-58-2 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65185-58-2
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD01529874
InChI Key ITRNQMJXZUWZQL-UHFFFAOYSA-N
Synonym 2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine
PubChem CID 2734091
IUPAC Name 2-(2-bromophenyl)ethanamine
SMILES C1=CC=C(C(=C1)CCN)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Refractive Index 1.5750 to 1.5770 (20°C, 589nm)
Quantity 5 g
Formula Weight 200.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromophenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only